Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 3306
GREENER_CHOICE

Catalog No. p-4816653
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-811-0 10 mg
41-815-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-811-0 Supplier Tocris Bioscience™ Supplier No. 4181/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cdk1 inhibitor

Ro 3306 is an ATP-competitive cyclin-dependent kinase 1 (cdk1) inhibitor (Ki values range from 35 to 240 nM depending on cdk1 binding partner). Also reported to inhibit other cdks (Ki values are 0.89-1.32, 0.03 and >2 μM for cdk2, cdk3 and cdk4, respectively). Induces G2/M phase cell cycle arrest and apoptosis. Downregulates the expression of antiapoptotic proteins such as Bcl-2 and survivin and enhances downstream p53 signaling in acute myeloid leukemia (AML). RO 3306 also improves homology-directed repair (HDR) -mediated gene editing in hematopoietic stem and progenitor cells.

Chemical Identifiers

CAS 872573-93-8
Molecular Formula C18H13N3OS2

Specifications

Quantity 10 mg
Formula Weight 351.45
Purity >98%
Molecular Weight (g/mol) 351.442
Chemical Name or Material Ro 3306
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.