Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ro 5-3335
GREENER_CHOICESDP

Catalog No. 469410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 46-941-0 Supplier R&D Systems™ Supplier No. 4694/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Core binding factor inhibitor; attenuates hematopoiesis

Ro 5-3335 is a core binding factor (CBF) inhibitor; preferentially kills human leukemia cell lines with CBF fusion proteins (IC50 = 1.1 μM). Represses RUNX1/CBFβ-dependent transactivation in reporter assays and inhibits transcriptional regulation by RUNX1 and CBFβ. Reduces leukemia burden in a mouse model. Attenuates RUNX1-dependent hematopoiesis in zebrafish embryos. Also a Tat antagonist; inhibits HIV-1 replication in vitro.

Specifications

Quantity 10 mg
Product Line Tocris
Product Type Ro 5-3335
Purity >98%
Molecular Weight (g/mol) 259.69
Inhibitors Ro 5-3335

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.