Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ro 60-0175 fumarate
GREENER_CHOICESDP

Catalog No. 185450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-545-0 50 mg
18-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-545-0 Supplier R&D Systems™ Supplier No. 1854/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective 5-HT2C agonist

Potent, selective 5-HT2 receptor agonist; shows selectivity for the 5-HT2C subtype (pKi values are 9, 7.5, 5.4, 5.2 and 5.6 for human 5-HT2C, 2A, 1A, 6 and 7 receptors respectively). Centrally active following oral or systemic administration in vivo.

Chemical Identifiers

CAS 169675-09-6
Molecular Formula C15H16ClFN2O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 342.75
Purity >98%
Molecular Weight (g/mol) 342.75
Chemical Name or Material Ro 60-0175 fumarate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.