Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ro 61-8048
GREENER_CHOICE

Catalog No. 325410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-541-0 10 mg
32-545-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-541-0 Supplier Tocris Bioscience™ Supplier No. 3254/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent kynurenine 3-monooxygenase (KMO) inhibitor

Ro 61-8048 is a potent and competitive kynurenine 3-monooxygenase (kynurenine 3-hydroxylase; KMO) inhibitor (Ki = 4.8 nM, IC50 = 37 nM). Increases kynurenic acid levels to concentrations that antagonize the glycine site of NMDA receptors. Brain penetrant and exhibits antidystonic, anticonvulsant and neuroprotective activities. Ro 61-8048 decreases nicotine self-administration in vivo. Ro 61-8048 prevents post-operative brain edema and consequent neuronal apoptosis in a rat model of surgically induced brain injury.

Chemical Identifiers

CAS 199666-03-0
Molecular Formula C17H15N3O6S2

Specifications

Quantity 10 mg
Formula Weight 421.45
Purity >99%
Molecular Weight (g/mol) 421.442
Chemical Name or Material Ro 61-8048
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.