Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ro 67-4853
GREENER_CHOICESDP

Catalog No. 434750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-475-0 50 mg
43-471-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-475-0 Supplier R&D Systems™ Supplier No. 4347/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Positive allosteric modulator of group I mGlu receptors

Positive allosteric modulator of mGlu group I receptors (pEC50 value is 7.16 for the rat mGlu1a receptor). Exhibits activity at all group I receptors, including human and rat mGlu1 and rat mGlu5. Enhances the effects of (S)-DHPG in CA3 neurons (EC50 = 95 nM).

Chemical Identifiers

CAS 302841-89-0
Molecular Formula C19H19NO4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 325.36
Purity >99%
Molecular Weight (g/mol) 325.364
Chemical Name or Material Ro 67-4853
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.