Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ro60 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501565801
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5801 Supplier Bethyl Laboratories, Inc Supplier No. A303693A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.

Specifications

Antigen Ro60
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene TROVE2
Gene Accession No. O08848, P10155
Gene Alias RP11-101E13.3, RO60, RORNP, SSA2, 60 kDa SS-A/Ro ribonucleoprotein, 60 kDa ribonucleoprotein Ro, Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro), gastric cancer multi-drug resistance protein, sjoegren syndrome type A antigen
Gene Symbols TROVE2
Host Species Rabbit
Immunogen Between 488 and 538
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20822, 6738
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.