Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROBLD3 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281696V
This item is not returnable.
View return policy
Description
ROBLD3 Polyclonal antibody specifically detects ROBLD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
ROBLD3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
ENDAP, Endosomal adaptor protein p14, HSPC003, Late endosomal/lysosomal adaptor and MAPK and MTOR activator 2, late endosomal/lysosomal adaptor, MAPK and MTOR activator 2, Late endosomal/lysosomal Mp1-interacting protein, MAPBPIPRoadblock domain-containing protein 3, MAPKSP1 adaptor protein, MAPKSP1AP, Mitogen-activated protein-binding protein-interacting protein, mitogen-activated protein-binding protein-interacting protein (MAPBPIP), p14Ragulator2, roadblock domain containing 3, ROBLD3 | |
ROBLD3 antibody was raised against an 18 amino acid peptide near the center of human ROBLD3. The immunogen is located within amino acids 40 - 90 of ROBLD3. Amino Acid Squence: WAAYDRNGNQAFNEDNLK | |
0.1 mL | |
Cancer | |
28956 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction