Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROBO1 Antibody (770502) [Janelia Fluor™ 646], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB71181J
Description
ROBO1 Monoclonal antibody specifically detects ROBO1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
ROBO1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
770502 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
Mouse myeloma cell line NS0-derived recombinant human ROBO1, Met1-Ala858, Accession # Q9Y6N7 | |
0.1 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
6091 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction