Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROBO1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323373
Description
ROBO1 Polyclonal antibody specifically detects ROBO1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
ROBO1 | |
Polyclonal | |
Western Blot 1:500, Immunocytochemistry/ Immunofluorescence 1:200 | |
DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) | |
Robo1 synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminal region of human Robo1. This sequence has significant homology to the same region in rat and mouse Robo1, and has low homology to other Robo family members. [UniProt# Q9Y6N7] | |
0.1 mL | |
Angiogenesis, Cancer, Extracellular Matrix | |
6091 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
PBS, 1 mg/mL BSA and 50% Glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction