Learn More
Specifications
Specifications
| Gene Accession No. | Q9Y6N7, Q9Y6N7, Q9Y6N7, Q9Y6N7 |
| Preparation Method | This product has been tested by SDS-PAGE and western blot and is suitable for use in Western blot, ELISA, Immunoprecipitation and ChIP. No detection of expected band at ~181 kDa is observed in ROBO1 knockout A549 when compared with unmodified A549 cell lysates by Western blot. |
| Quantity | 100 μg |
| Purity | Multi-step |
| Host Species | Human |
| Content And Storage | Store at -70°C. Avoid freeze/thaw cycles. |
| Product Line | Novus |
| Research Category | Angiogenesis, Cancer, Extracellular Matrix |
| Target | ROBO1 |
| Synonym | DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
