Learn More
Description
- Gene name: ROBO1
- Gene Alias: DUTT1, FLJ21882, MGC131599, MGC133277, SAX3
- Entrez GeneID: 6091
- strong>Gene Description: Roundabout, axon guidance receptor, homolog 1
Specifications
Specifications
| Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
| Description | Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for 2 transcript variants. (NM_001145845.1, NM_133631.3) |
| Species | Human |
| For Use With (Application) | RNAi Knockdown |
| Includes | DEPC water |
| Product Type | ROBO1 Pre-design Chimera RNAi |
| Quantity | 10 nmol |
| Target | Human |
For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.