Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROBO2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$416.50 - $670.00
Specifications
Antigen | ROBO2 |
---|---|
Concentration | 0.05mg/mL |
Dilution | Western Blot Reported in scientific literature (PMID: 32220420)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated Reported in scientific literature (PMID: 32220420). |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay |
Classification | Polyclonal |
Description
ROBO2 Polyclonal specifically detects ROBO2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
ROBO2 | |
Western Blot Reported in scientific literature (PMID: 32220420)., Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500, Knockout Validated Reported in scientific literature (PMID: 32220420). | |
Polyclonal | |
Rabbit | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
6092 | |
This antibody was developed against Recombinant ROBO2 Protein corresponding to amino acids: PVNNSNSGPNEIGNFGRGDVLPPVPGQGDKTATMLSDGAIYSSIDFTTKTSYNSSSQITQATPYATTQILHSNSIHELAVDLPDPQWKSSIQQKTDLMGFGYSLPDQNKGNNGGKGGKKKKNKNSSKPQKNNGSTWA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.05mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoassay | |
Unconjugated | |
RUO | |
Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila) | |
ROBO2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title