Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROBO2 Monoclonal Antibody (356001)
GREENER_CHOICE

Catalog No. PIMA524104
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524104 Supplier Invitrogen™ Supplier No. MA524104
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ROBO1, rhROBO3, or rhROBO4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

ROBO2 belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. ROBO2 is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for ROBO2.This gene belongs to the ROBO family, part of the immunoglobulin superfamily proteins that are highly conserved from fly to human. The encoded protein is a receptor for SLIT2, molecules known to function in axon guidance and cell migration. Defects in this gene are the cause of vesicoureteral reflux type 2. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROBO2
Applications Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 356001
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ROBO2
Gene Accession No. Q9HCK4
Gene Alias 2600013A04Rik; 9430089E08Rik; ast; astray; BB097918; D230004I22Rik; KIAA1568; mKIAA1568; ROBO2; roundabout 2; roundabout guidance receptor 2; roundabout homolog 2; roundabout homolog 2 (Drosophila); roundabout homolog 2-like; roundabout, axon guidance receptor, homolog 2; roundabout, axon guidance receptor, homolog 2 (Drosophila); roundabout2; SAX3; sb:eu937; Unknown (protein for IMAGE:8049638); unp1881; zmp:0000000549
Gene Symbols ROBO2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human ROBO2 isoform 1 Ser22-Pro859.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6092
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.