Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ ROBO2 Pre-design Chimera RNAi
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variants mRNA. This product targets for all of 5 transcript variants (NM_001128929,NM_001290039,NM_001290040,NM_001290065,NM_002942)
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
Species | Human |
For Use With (Application) | RNAi Knockdown |
Includes | DEPC water |
Product Type | ROBO2 Pre-design Chimera RNAi |
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Price | Quantity & Availability | ||||
89-035-718
|
Abnova™
H00006092R03S |
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variants mRNA. This product targets for all of 5 transcript variants. (NM_001128929,NM_001290039,NM_001290040,NM_001290065,NM_002942) | 10 nmol | N/A | N/A | N/A | |||
89-035-720
|
Abnova™
H00006092R04L |
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variant 4, mRNA. | 20 nmol | N/A | N/A | N/A | |||
89-035-719
|
Abnova™
H00006092R03L |
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variants mRNA. This product targets for all of 5 transcript variants. (NM_001128929,NM_001290039,NM_001290040,NM_001290065,NM_002942) | 20 nmol | N/A | N/A | N/A | |||
89-035-721
|
Abnova™
H00006092R04S |
Homo sapiens roundabout, axon guidance receptor, homolog 2 (Drosophila) (ROBO2), transcript variant 4, mRNA. | 10 nmol | N/A | N/A | N/A | |||
Description
- Gene name: ROBO2
- Gene Alias: KIAA1568,SAX3
- Entrez GeneID: 6092
- Gene Description: Roundabout, axon guidance receptor, homolog 2 (Drosophila)
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
RNAi Knockdown | |
ROBO2 Pre-design Chimera RNAi |
Human | |
DEPC water | |
Human |
For research use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title