Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Robo3 Rabbit anti-Bovine, Human, Non-human Primate, Panda, Polyclonal, FabGennix

Catalog No. 502208855
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8855 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8855 Supplier Fabgennix Inc Supplier No. RIG1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Robo3 Polyclonal antibody specifically detects Robo3 in Bovine, Human, Non-human Primate, Panda samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen Robo3
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Robo3
Gene Accession No. Q96MS0
Gene Alias FLJ21044; HGPPS; HGPS; RBIG1; retinoblastoma inhibiting gene 1; retinoblastoma-inhibiting gene 1 protein; Rig1; Rig-1; ROBO3; Robo3a; Robo3b; roundabout guidance receptor 3; roundabout homolog 3; roundabout homolog 3 (Drosophila); roundabout, axon guidance receptor, homolog 3; roundabout, axon guidance receptor, homolog 3 (Drosophila); roundabout-like protein 3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to unique amino acid sequence on human RIG1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100337359, 100469109, 466839, 64221
Target Species Bovine, Human, Non-human Primate, Panda
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.