Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROBO4 Antibody (003), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290031R
Description
ROBO4 Monoclonal specifically detects ROBO4 in Human samples. It is validated for ELISA.Specifications
ROBO4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
3 | |
ELISA | |
ECSM4, FLJ20798, Magic roundabout, MGC133352, MGC133353, MRB, roundabout homolog 4, roundabout homolog 4, magic roundabout (Drosophila) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ROBO4 (Q8WZ75-1; Met1-Glu469). | |
0.1 mL | |
Angiogenesis, Cancer, Extracellular Matrix, Neuroscience | |
54538 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction