Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ROBO4 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547304
Description
In direct ELISAs, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO2, and rhROBO3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins. Involved in the maintenance of endothelial barrier organization and function (PubMed:30455415). [UniProt].
Specifications
| ROBO4 | |
| Polyclonal | |
| Unconjugated | |
| ROBO4 | |
| 1200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO3674 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 54538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8WZ75 | |
| ROBO4 | |
| Mouse myeloma cell line NS0-derived recombinant human ROBO4 Gln28-Arg467. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction