Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROBO4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597279 Supplier Invitrogen™ Supplier No. PA597279
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVGPKGGVLL CPPRPCLTPT PSEGSLANGW GSASEDNAAS ARASLVSSSD GSFLADAHFA RALAVAVDSF GFGLEPREAD CVFIDASSPP SPRDEIFLTP NLSLPLWEWR PDWLEDMEVS HTQRLGRGMP PWPPDSQISS QRSQLHCRMP KAGASPVDYS; Positive Samples: HT-29, HepG2, HT-1080, HeLa.

Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins. Involved in the maintenance of endothelial barrier organization and function (PubMed:30455415). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROBO4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ROBO4
Gene Accession No. Q8WZ75
Gene Alias 1200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO3674
Gene Symbols ROBO4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 848-1007 of human ROBO4 (NP_0619284).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54538
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.