Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROBO4 Recombinant Rabbit Monoclonal Antibody (036)
GREENER_CHOICE

Catalog No. PIMA531222
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531222 Supplier Invitrogen™ Supplier No. MA531222
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse ROBO4.

Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins. Involved in the maintenance of endothelial barrier organization and function (PubMed:30455415). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROBO4
Applications ELISA
Classification Recombinant Monoclonal
Clone 036
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ROBO4
Gene Accession No. Q8C310
Gene Alias 1200012D01Rik; AI593217; ECSM4; FLJ20798; Magic roundabout; MGC133352; MGC133353; MRB; ROBO4; roundabout guidance receptor 4; roundabout homolog 4; roundabout homolog 4 (Drosophila); roundabout homolog 4, magic roundabout; roundabout, axon guidance receptor, homolog 4; roundabout, axon guidance receptor, homolog 4 (Drosophila); UNQ421/PRO3674
Gene Symbols ROBO4
Host Species Rabbit
Immunogen Recombinant Mouse ROBO4 protein (Met1-Glu480).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74144
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.