Learn More
Stoelting™ Robot and Motorized Stereotaxic Accessories
Innovative accessories for use with the Robot and Motorized Stereotaxic Systems
Supplier: Stoelting™ 51709
Description
- Provides automated drilling followed by an intracranial injection using the same holder
- Ideal for multiple injection sites with no tool exchange between the drill and injector
- Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
- Available as Glass-Capillary Nanoinjector
- Atlas Integration
- High Throughput Drilling and Injection
- No Tool Exchange
- Defined/Stored Protocols
- Multisite Microinjections
- Ideal for Optogenetics applications
- Precise Depth Control
- Brain Windowing
- Contour Drilling
- Smooth Edges
- Skull Thinning
Specifications
Neuroscience | |
Mouse | |
Microinjector Robot System |