Learn More
Stoelting™ Robot and Motorized Stereotaxic Accessories
SDP
Innovative accessories for use with the Robot and Motorized Stereotaxic Systems
Specifications
For Use With (Application) Neuroscience
Animal Mouse
View More Specs
Products 5
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
10-000-193
SDP
View Documents
Stoelting™
51708
Optogenetics Robot System
Each for $72,550.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
10-000-194
SDP
View Documents
Stoelting™
51709
Microinjector Robot System
Each for $48,700.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
10-000-195
SDP
View Documents
Stoelting™
51710
Robot Drill System
Each for $48,700.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
10-000-196
SDP
View Documents
Stoelting™
51711
Drill/Capillary Nanoinjection Robot
Each for $63,500.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
10-000-197
SDP
View Documents
Stoelting™
51712
Capillary Nanoinjection Robot
Each for $48,700.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Provides automated drilling followed by an intracranial injection using the same holder
  • Ideal for multiple injection sites with no tool exchange between the drill and injector
  • Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
  • Available as Glass-Capillary Nanoinjector
  • Atlas Integration
  • High Throughput Drilling and Injection
  • No Tool Exchange
  • Defined/Stored Protocols
  • Multisite Microinjections
  • Ideal for Optogenetics applications
  • Precise Depth Control
  • Brain Windowing
  • Contour Drilling
  • Smooth Edges
  • Skull Thinning
Specifications
Specifications
Neuroscience
Mouse
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.