Learn More
Stoelting™ Robot and Motorized Stereotaxic Accessories
SDP
Innovative accessories for use with the Robot and Motorized Stereotaxic Systems
Supplier:  Stoelting™ 51712
 View more versions of this product
Catalog No. 10-000-197
Only null left
Add to Cart
Request a Quote
Description
Description
  • Provides automated drilling followed by an intracranial injection using the same holder
  • Ideal for multiple injection sites with no tool exchange between the drill and injector
  • Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
  • Available as Glass-Capillary Nanoinjector
  • Atlas Integration
  • High Throughput Drilling and Injection
  • No Tool Exchange
  • Defined/Stored Protocols
  • Multisite Microinjections
  • Ideal for Optogenetics applications
  • Precise Depth Control
  • Brain Windowing
  • Contour Drilling
  • Smooth Edges
  • Skull Thinning
Specifications
Specifications
Neuroscience
Mouse
Capillary Nanoinjection Robot
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.