Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIOTIX™ Robotic aTIP 70μL Filter Pipette Tips
GREENER_CHOICE

Catalog No. 12111062
Click to view available options
Volume (Metric):
70 μL
This item is not returnable. View return policy
This item is not returnable. View return policy

For use with Agilent™ Velocity 11™ VPrep and Bravo, 384-Head platforms.

  • Made from clear polypropylene
  • Compatible with 96-and 384-well plates
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 70 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Compatibility Agilent/Velocity 11 Automation Systems
Format Lift-off Lid Rack
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Agilent/Velocity 11 VPrep and Bravo, 384 Tip Format
Graduated No
Material Polypropylene
Shape Conical
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.