Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110664 Supplier Invitrogen™ Supplier No. PA5110664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAAMDVDTP SGTNSGAGKK RFEVKKWNAV ALWAWDIVVD NCAICRNHIM DLCIECQANQ ASATSEECTV AWGVCNHAFH FHCISRWLKT RQVCPLDNRE WEF Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROC1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBX1
Gene Accession No. P62877
Gene Alias 1500002P15Rik; AA517855; BA554C12.1; E3 ubiquitin-protein ligase RBX1; E3 ubiquitin-protein ligase RBX1, N-terminally processed; E3 ubiquitin-protein transferase RBX1; E3 ubiquitin-protein transferase RBX1, N-terminally processed; Protein ZYP; Rbx1; Regulator of cullins 1; RING box protein 1; RING finger protein 75; ring-box 1; ring-box 1, E3 ubiquitin protein ligase; RING-box protein 1; RNF75; ROC1; ZYP protein
Gene Symbols RBX1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human ROC1. Recombinant protein control fragment (Product #RP-91505).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9978
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.