Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Roche Diagnostics Accutrend™ Plus Meter Kit
Shop All Roche Diagnostics Products
Click to view available options
Quantity:
Each
Description
- Prevent cross-contamination and improve patient safety with out-of-meter dosing
- Fast, reliable results
- Lightweight, ergonomic design makes meter easy to handle and operate
- Large, easy-to-read screen helps user view results and interpret them accurately
- Built-in performance quality control testing uses multiple checks to ensure accurate and reliable results
- Positive identification of strip lot and code parameters helps ensure accurate results and proper system performance
- Meter stores up to 100 results for both test parameters (cholesterol)
Specifications
Specifications
| Sample Type | Fresh Capillary Blood |
| Type | Accutrend Plus™ Meter Kit |
| For Use With (Application) | Monitoring cholesterol |
| Includes | Four AAA batteries |
| Memory | 100 Values for each parameter |
| Quantity | Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction