Learn More
Roche Diagnostics CoaguChek™ XS Plus System
Catalog No. p-3984930 Shop All Roche Diagnostics Products
Click to view available options
Type:
CoaguChek Handheld Base Unit
XS Plus System
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
CoaguChek XS Plus instrument, handheld power supply unit, printed manual in English, CD-ROM with other languages: German, French, Italian, English, Spanish, Portuguese, Danish, Norwegian, Swedish, Finnish and Dutch
Requires
PT strips (in a vial): Box with 2 vials of 24 strips each with a code chip; Box with 1 vial of 24 strips with a code chip.
Built-in QC and data management for POC anticoagulation testing
The professional solution for clinical practices.
Accurate and flexible technology — the smart way to test
  • Accurate, precise results in about one minute
  • The only system to perform onboard quality control and determine patient results in a single test chamber
  • Neutralizes therapeutic levels of heparin and LMWH
  • INR corrected for hematocrit within specified range
Streamlined data management
  • Stores 1000 patient and 500 QC test results
  • Operator and QC lockout capabilities
  • Code chip programming
  • Data storage for up to 60 code chips
Easy and convenient to use
  • Small sample size (8μL) from a fingerstick
  • Top or side strip dosing, with outside-meter blood application (avoiding potential cross-contamination)
  • New onboard controls — no external QC necessary
  • On-the-spot adjustment of your patient's VKA dosage — no need to wait for lab results
TRUSTED_SUSTAINABILITY
Specifications
Type XS Plus System
Includes CoaguChek XS Plus instrument, handheld power supply unit, printed manual in English, CD-ROM with other languages: German, French, Italian, English, Spanish, Portuguese, Danish, Norwegian, Swedish, Finnish and Dutch
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.