Learn More
Roche Diagnostics CoaguChek™ XS System
Catalog No. 22-045-960 Shop All Roche Diagnostics Products
Click to view available options
:
Coaguchek XS System
Accurate and flexible technology
The CoaguChek XS System uses exclusive smart technology to help ensure accurate results offering the flexibility and control you need to confidently manage a wide range of anticoagulated patients.
  • Accurate, precise results in one minute with a fingerstick test
  • The only system to perform onboard quality control in the reagent chamber† ‡
  • Neutralizes therapeutic levels of heparin and LMWH†
  • INR corrected for hematocrit within specified range – manage a broad array of patients with one device†
TRUSTED_SUSTAINABILITY
Specifications
Type CoaguChek XS System
For Use With (Application) For monitoring oral anticoagulation therapy
Includes Meter, batteries, user manual and carrying case
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.