Learn More
Roche Diagnostics LightCycler™ Capillaries
Reaction vessels for LightCycler™ Carousel-Based System Instruments
Supplier: Roche Diagnostics 04929292001

Description
A glass capillary is molded to a polypropylene reservoir, facilitating easy loading.
- Stopper provides a secure seal, significantly reducing the risk of contamination
- High-quality borosilicate glass ensures superior PCR performance and optimal fluorescence transmission
- The capillary can hold reaction volumes ranging from 10 to 20 μL

Specifications
LightCycler™ Carousel-Based System Instruments | |
Reaction Vessels | |
LightCycler™ Capillaries |
Fluorescence | |
PCR | |
10 to 20 μL |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.