Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK1 Antibody (154C1465), DyLight 755 - (active/cleaved), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10056596IR
Description
ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse, Rat, Canine, Primate, Rabbit samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
cleaved ROCK1 | |
Monoclonal | |
DyLight 755 | |
O60259 | |
ROCK1 | |
This antibody was generated by immunizing mice with a synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1. This sequence is 100% conserved between human, chimpanzee, mouse, rat, dog, and rabbit. This immunogen peptide sequence is adjacent to the caspase-3 cleavage site of ROCK-1. | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
6093 | |
Human, Mouse, Rat, Canine, Primate, Rabbit | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
154C1465 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for detection of apoptosis. | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction