Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK1 Antibody (261), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242917
Description
Ensure accurate, reproducible results in Western Blot
ROCK1 Monoclonal specifically detects ROCK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ROCK1 | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
ROCK1 | |
Recombinant protein encompassing a sequence within the N-terminus region of human ROCK1. The exact sequence is proprietary. | |
0.1 mL | |
Apoptosis, Breast Cancer, Cancer, Signal Transduction, Wnt Signaling Pathway | |
6093 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
261 | |
Western Blot 1:500-1:3000 | |
EC 2.7.11, EC 2.7.11.1, MGC131603, MGC43611, p160 ROCK-1, p160ROCK, p160-ROCK, PRO0435, Renal carcinoma antigen NY-REN-35, Rho kinase, rho-associated protein kinase 1, Rho-associated, coiled-coil containing protein kinase 1, Rho-associated, coiled-coil-containing protein kinase 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction