Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROCK1 (Cleaved Asp1113) Monoclonal Antibody (154C1465)
GREENER_CHOICE

Catalog No. 01676444
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-444 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-444 Supplier Invitrogen™ Supplier No. MA141031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mAb recognizes the cleaved C-terminus of ROCK-1. It does not recognize the full-length protein. Since ROCK-1 is cleaved by caspase-3 during apoptosis, this may be used for the detection of apoptosis. A suggested positive control is Jurkat cells treated with camptothecin or anti-Fas antibody to induce apoptosis. Cross reacts with Chimpanzee.

The Rho target protein, Rho-associated coiled coil-containing protein kinase (ROCK)-1 has been found to be a new Caspase-3 substrate. ROCK, one of the effectors of the small GTPase Rho, has recently been shown to contribute significantly to myosin light chain (MLC) activation through two pathways: direct phosphorylation of MLC and phosphorylation of MLC phosphatase, leading to its inhibition. The increase in cellular contractility that is necessary for apoptotic membrane blebbing implies sustained augmentation of MLC phosphorylation. ROCK-1 consists of an amino-terminal kinase domain and an inhibitory cysteine/histidine-rich C-terminal domain that is located within a pleckstrin-homology region. They are joined by a variable region that contains the Rho-binding domain. During apoptosis, ROCK-I is cleaved by caspase-3 at a conserved DETD1113/G sequence and its carboxy-terminal inhibitory domain is removed, resulting in deregulated and constitutive kinase activity. The caspase-3-mediated cleavage and activation of ROCK I induces phosphorylation of MLC and membrane blebbing, one of the first events in the execution phase of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROCK1 (Cleaved Asp1113)
Applications Western Blot
Classification Monoclonal
Clone 154C1465
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ROCK1
Gene Accession No. O77819, P70335, Q13464, Q63644
Gene Alias 1110055K06Rik; Ac2-154; cAMP-dependent protein kinase ROCK-I; cePKA; corneal epithelial Rho-associated-ser/thr kinase; corneal epithelial Rho-associated-Ser/Thr kinase 1; HEBM1; liver regeneration-related protein LRRG199; MGC131603; MGC43611; p150 RhoA-binding kinase ROK beta; p160 ROCK-1; p160ROCK; PRO0435; renal carcinoma antigen NY-REN-35; Rho associated coiled-coil containing protein kinase 1; Rho-associated coiled-coil containing protein kinase 1; Rho-associated coiled-coil forming kinase 1; Rho-associated kinase beta; rho-associated protein kinase 1; Rho-associated, coiled-coil containing protein kinase 1; rho-associated, coiled-coil-containing protein kinase 1; Rho-associated, coiled-coil-containing protein kinase I; ROCK1; Rock-I
Gene Symbols ROCK1
Host Species Mouse
Immunogen Synthetic peptide sequence corresponding to amino acids 1114-1126 GNLPESRIEGWLS of human ROCK-1.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009220, 19877, 480181, 6093, 81762
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.