Learn More
Promega ROCK1 Kinase Enzyme System, 1mg
Catalog No. PRV3412 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3412 1 mg
1 options
Catalog No. PRV3412 Supplier Promega Supplier No. V3412
Add to Cart
Add to Cart
Request a Quote
Recombinant human ROCK1 (amino acids 17-535) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ROCK1 is a ubiquitously expressed serine-threonine kinase that is a downstream target of the small GTPase RhoA.
  • Easily Screen and Profile ROCK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C