Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROCK1/ROCK2 Recombinant Rabbit Monoclonal Antibody (HL1632)
GREENER_CHOICE

Catalog No. FEMA550324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550324 Supplier Invitrogen™ Supplier No. MA550324
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ROCK1/ROCK2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

ROCK1 and ROCK2 are genes that encode for Rho-associated coiled-coil containing protein kinases, also known as ROCKs. These kinases are involved in a variety of cellular processes such as cell migration, adhesion, proliferation, and apoptosis. They are activated by the small GTPase RhoA and phosphorylate downstream targets such as myosin light chain, LIM kinase, and myosin phosphatase. Dysregulation of ROCK1 and ROCK2 has been linked to various diseases including cancer, cardiovascular disease, and neurological disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROCK1/ROCK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1632
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ROCK2
Gene Accession No. O75116, Q13464
Gene Alias ROCK-1/ROCK-2; ROCK-I/ROCK-II
Gene Symbols ROCK1, ROCK2
Host Species Rabbit
Immunogen Recombinant fragment of human ROCK1 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6093, 9475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.