Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK2 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100593AF750
Description
ROCK2 Polyclonal antibody specifically detects ROCK2 in Human samples. It is validated for Western BlotSpecifications
ROCK2 | |
Polyclonal | |
Western Blot | |
coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2 | |
The immunogen recognized by this antibody maps to a region between residues 1320 and the C-terminus (residues 1388) of human ROCK2 using the number given in entry NP_004841.1 (GeneID 9475) | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific, Protein Kinase, Wnt Signaling Pathway | |
9475 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction