Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK2 Antibody [Janelia Fluor™ 549], Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals AF4790I
Description
ROCK2 Polyclonal antibody specifically detects ROCK2 in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
ROCK2 | |
Polyclonal | |
50mM Sodium Borate | |
Goat | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2 | |
E. coli-derived recombinant human ROCK2, Ile641-Ala846, Accession # O75116 | |
0.1 mL | |
Breast Cancer, Cancer, Phospho Specific, Protein Kinase, Wnt Signaling Pathway | |
9475 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction