Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROCK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100593
Description
ROCK2 Polyclonal specifically detects ROCK2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
ROCK2 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
coiled-coil-containing protein kinase 2, EC 2.7.11, EC 2.7.11.1, KIAA0619, p164 ROCK-2, Rho kinase 2, rho-associated protein kinase 2, Rho-associated, coiled-coil containing protein kinase 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB100-593 is specific for human Rho Kinase alpha protein. | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:10000-1:25000, Immunoprecipitation 2-5 ug/mg lysate | |
O75116 | |
ROCK2 | |
The immunogen recognized by this antibody maps to a region between residues 1320 and the C-terminus (residues 1388) of human ROCK2 using the number given in entry NP_004841.1 (GeneID 9475) | |
100 μL | |
Breast Cancer, Cancer, Protein Kinase, Wnt Signaling Pathway | |
9475 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction