Learn More
Promega ROCK2 Kinase Enzyme System, 1mg
Catalog No. PRV6479 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6479 1 mg
1 options
Catalog No. PRV6479 Supplier Promega Supplier No. V6479
Add to Cart
Add to Cart
Request a Quote
ROCK2 is a ubiquitously expressed serine/threonine kinase localized in the nucleus that regulates cytokinesis, smooth muscle contraction, the formation of actin stress fibers and focal adhesions, and the activation of the c-fos serum response element.
  • Easily Screen and Profile ROCK2 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C