Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROCK2 Mouse anti-Human, Mouse, Rat, Clone: 1C7B8, Proteintech
SDP

Catalog No. 501737051 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7051 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7051 Supplier Proteintech Group Inc Supplier No. 666331IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specific to ROCK2.

The function of this protein remains unknown.

Specifications

Antigen ROCK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1C7B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ROCK2
Gene Accession No. O75116, P70336, Q62868
Gene Alias KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2
Gene Symbols ROCK2
Host Species Mouse
Immunogen ROCK2 Fusion Protein Ag16330
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19878, 25537, 9475
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.