Learn More
ROD/KNTC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301713AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.Specifications
| ROD/KNTC1 | |
| Polyclonal | |
| Unconjugated | |
| KNTC1 | |
| Kinetochore associated 1, FLJ36151, KIAA0166, ROD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9735 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| P50748 | |
| KNTC1 | |
| Between 2159 and 2209 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.