Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ROD1 (PTBP3) Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene
SDP

Catalog No. 502543835
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3835 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3835 Supplier OriGene Supplier No. CF812036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROD1 (PTBP3) Monoclonal antibody specifically detects ROD1 (PTBP3) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ROD1 (PTBP3)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C9
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene Sh3bgr
Gene Accession No. P55822
Gene Alias 5830471K22Rik; AA407443; AI462022; AW107884; C86549; fission yeast differentiation regulator; polypyrimidine tract binding protein 3; polypyrimidine tract-binding protein 3; PTBP3; regulator of differentiation (in S. pombe) 1; regulator of differentiation 1; regulator of differentiation 1; polypyrimidine tract-binding protein 3; ROD1; ROD1 regulator of differentiation 1; si:dkey-27p1.2
Host Species Mouse
Immunogen Full length human recombinant protein of human SH3BGR (NP_031367) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6450
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.