Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROGDI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546617 Supplier Invitrogen™ Supplier No. PA546617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VHAVLKQLQD ILKEASLRFT LPGSGTEGPA KQENFILGSC GTDQVKGVLT Sequence homology: Cow: 93%; Dog: 81%; Guinea Pig: 93%; Human: 100%; Mouse: 86%; Rabbit: 86%; Rat: 93%.

This gene encodes a protein of unknown function. Loss-of-function mutation in this gene cause Kohlschutter-Tonz syndrome. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROGDI
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ROGDI
Gene Accession No. Q9GZN7
Gene Alias 0610011C19Rik; AU020118; C76152; KTZS; leucine zipper domain protein; leucine-zipper-containing LZF; Lzf; protein rogdi homolog; ROGDI; rogdi atypical leucine zipper; rogdi homolog; rogdi homolog (Drosophila)
Gene Symbols ROGDI
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ROGDI (aa 27-76).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79641
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.