Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROGDI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597197 Supplier Invitrogen™ Supplier No. PA597197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATVMAATAA ERAVLEEEFR WLLHDEVHAV LKQLQDILKE ASLRFTLPGS GTEGPAKQEN FILGSCGTDQ VKGVLTLQGD ALSQADVNLK MPRNNQLLHF AFREDKQWKL QQIQDARNHV SQAIYLLTSR DQSYQFKTGA; Positive Samples: Mouse brain, Rat brain; Cellular Location: Nucleus envelope.

This gene encodes a protein of unknown function. Loss-of-function mutation in this gene cause Kohlschutter-Tonz syndrome. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROGDI
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ROGDI
Gene Accession No. Q3TDK6, Q4V7D2
Gene Alias 0610011C19Rik; AU020118; C76152; KTZS; leucine zipper domain protein; leucine-zipper-containing LZF; Lzf; protein rogdi homolog; ROGDI; rogdi atypical leucine zipper; rogdi homolog; rogdi homolog (Drosophila)
Gene Symbols ROGDI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human ROGDI (NP_0788651).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 287061, 66049
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.