Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Rolipram
GREENER_CHOICESDP

Catalog No. 090550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-055-0 50 mg
09-051-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-055-0 Supplier R&D Systems™ Supplier No. 0905/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. PDE4 inhibitor

Rolipram is a selective inhibitor of cAMP phosphodiesterase (PDE4) (IC50 = 2.0 μM). Discriminates between two conformational states of PDE4 isoenzymes.

R-enantiomer also available.

Chemical Identifiers

CAS 61413-54-5
Molecular Formula C16H21NO3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 275.35
Purity >98%
Molecular Weight (g/mol) 275.35
Chemical Name or Material Rolipram
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.