Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Rolipram, 98%

Catalog No. AAJ67177MB
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67177MB 25 mg
AAJ67177LB0 5 mg
AAJ67177MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67177MB Supplier Thermo Scientific Chemicals Supplier No. J67177MB

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61413-54-5
Molecular Formula C16H21NO3
Synonym 4-(3-Cyclopentyloxy-4-methoxyphenyl)-2-pyrrolidinone; ZK 62711

Specifications

Quantity 25 mg
Description White to off-white
Content And Storage Ambient temperatures
Formula Weight 275.35g/mol
Form Solid
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.