Learn More
Rolling Collision Balls
Catalog No. s40973
Click to view available options
:
Balls Collision Glass Set 6/pk
Track and collision balls demonstrate actions and reactions.
  • Demonstrate the principle that for every action there is an equal and opposite reaction
  • Include a pair of plated steel tracks 31.9 in. (81cm) long and seven steel collision balls 1 in. (2.5cm) in diameter
  • An optional set of six glass collision balls is also available (does not include track)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.