Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565033 Supplier Invitrogen™ Supplier No. PA565033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHYKDTEVPGH CQAKRLVDEL QLRYHCCGRH GYKDWFGVQW VSSRYLDPGD RDVADRIQSN VEGLYLTDGV PFSCCNPHSP RPCLQNRLSD SYAHPLFDPR QPNQNLWA Highest antigen sequence identity to the following orthologs - mouse 90%, rat 92%.

This gene is a member of a photoreceptor-specific gene family and encodes an integral membrane protein found in the photoreceptor disk rim of the eye. This protein can form homodimers or can heterodimerize with another photoreceptor, retinal degeneration slow (RDS). It is essential for disk morphogenesis, and may also function as an adhesion molecule involved in the stabilization and compaction of outer segment disks or in the maintenance of the curvature of the rim. Certain defects in this gene have been associated with the degenerative eye disease retinitis pigmentosa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ROM1
Gene Accession No. Q03395, Q5PPM7
Gene Alias M101156; retinal outer segment membrane protein 1; Rgsc1156; Rod outer segment membrane protein 1; ROM; Rom1; Rom-1; ROSP1; RP7; Tetraspanin-23; TSPAN23; tspan-23
Gene Symbols ROM1
Host Species Rabbit
Immunogen Recombinant Human ROM1. Recombinant protein control fragment (Product #RP-105799).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309201, 6094
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.