Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ROMO1 Monoclonal Antibody (OTI1C7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505583S


Description
ROMO1 Monoclonal antibody specifically detects ROMO1 in Human samples. It is validated for Western BlotSpecifications
ROMO1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P60602 | |
Mouse | |
Affinity chromatography | |
RUO | |
140823 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI1C7 | |
Unconjugated | |
ROMO1 | |
2010100O12Rik; AI853864; bA353C18.2; C13H20ORF52; C20orf52; Epididymis tissue protein Li 175; glyrichin; hypothetical protein MGC73345; mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; MTGMP; PCM19; protein MGR2 homolog; RA_r333_jsm669B4f; Reactive oxygen species modulator 1; romo1; ROS modulator 1; Unknown (protein for MGC:127858); zgc:73345; zgc:77111 | |
Full length human recombinant protein of human ROMO1 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction