Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ROMO1 Monoclonal Antibody (OTI2C12)
GREENER_CHOICE

Catalog No. PIMA526167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526167 Supplier Invitrogen™ Supplier No. MA526167
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ROMO1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The protein encoded by this gene is a mitochondrial membrane protein that is responsible for increasing the level of reactive oxygen species (ROS) in cells. The protein also has antimicrobial activity against a variety of bacteria by inducing bacterial membrane breakage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ROMO1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ROMO1
Gene Accession No. P60602
Gene Alias 2010100O12Rik; AI853864; bA353C18.2; C13H20ORF52; C20orf52; Epididymis tissue protein Li 175; glyrichin; hypothetical protein MGC73345; mitochondrial targeting GxxxG motif protein; mitochondrial targeting GXXXG protein; MTGM; MTGMP; PCM19; protein MGR2 homolog; RA_r333_jsm669B4f; Reactive oxygen species modulator 1; romo1; ROS modulator 1; Unknown (protein for MGC:127858); zgc:73345; zgc:77111
Gene Symbols ROMO1
Host Species Mouse
Immunogen Full length human recombinant protein of ROMO1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140823
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.