Learn More
Promega RON Kinase Enzyme System, 1mg
Catalog No. PRV3922 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV3922 1 mg
1 options
Catalog No. PRV3922 Supplier Promega Supplier No. V3922
Add to Cart
Add to Cart
Request a Quote
Recombinant human RON (amino acids 983-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RON is a macrophage stimulating 1 receptor c-met-related tyrosine kinase.
  • Easily Screen and Profile RON Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C