Learn More
RON Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | RON |
---|---|
Concentration | 1.06 mg/mL |
Applications | ELISA, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2289
![]() ![]() |
Rockland Immunochemicals
600401K07 |
100 μg |
Each for $636.24
|
|
|||||
50-199-2290
![]() ![]() |
Rockland Immunochemicals
600401K07S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum. This antibody is specific for human RON. Cross-reactivity against RON from other species may occur but has not yet been tested.
Macrophage-stimulatin 1 receptor (MST1R/Ron), a HGF Receptor/MET-type protein kinase, mediates the biological activities of macrophage-stimulating protein (MSP), a multifunctional cytokine that regulates cell adhesion, motility, growth, and survival. The protein is a membrane-spanning, disulfide-linked heterodimer, which results from cleavage of a glycosylated precursor into 35-kD (alpha) and 150-kD (beta) subunits. Ligand binding results in tyrosine phosphorylation of the beta chain. In knockout studies, MST1R/RON (-/-) mice failed to survive past the periimplantation period. The MST1R/RON gene has been mapped to 3p21, a region of frequent deletion or mutation in small cell lung and renal carcinoma, and has been implicated in the progression of several epithelial cancers. MST1R/Ron expression has been documented in many normal human tissues. ESTs have been isolated from several tissue libraries, including normal colon, mouth, prostate, and testis and cancerous colon, prostate, stomach, and uteru.Specifications
RON | |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q04912 | |
4486 | |
Primary | |
Antibody |
1.06 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
CD136; CDw136; c-met-related tyrosine kinase; friend virus susceptibility 2; Fv2; Fv-2; macrophage stimulating 1 receptor; macrophage stimulating 1 receptor (c-met-related tyrosine kinase); Macrophage-stimulating protein receptor; Macrophage-stimulating protein receptor alpha chain; Macrophage-stimulating protein receptor beta chain; MSP receptor; Mst1r; MST1R variant RON30; MST1R variant RON62; p185-Ron; protein tyrosine kinase 8; Protein-tyrosine kinase 8; PTK8; PTK8 protein tyrosine kinase 8; receptor protein tyrosine kinase, c-met-related; RON; RON variant E2E3; Stem cell-derived tyrosine kinase; Stk | |
MST1R | |
-20°C, Avoid Freeze/Thaw Cycles | |
MST1R |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.